Learn More
Salbutamol hemisulfate, MedChemExpress
Product Code. 16458306
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16458306
Brand: Medchem Express HYB0436
MedChemExpress Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting β2 adrenergic receptor agonist Target: β2 Adrenergic Receptor Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting, selective beta2-adrenergic receptor agonist used in the treatment of asthma and COPD.
Salbutamol Hemisulfate (Albuterol hemisulfate) is a short-acting β2 adrenergic receptor agonist
Specifications
Chemical Name or Material Salbutamol hemisulfate
CAS 51022-70-9
Color White
Purity Grade Notes Research
Molecular Formula C13H22NO5S0.5
Quantity 100 mg
Synonym Albuterol hemisulfateAH-3365 hemisulfate
Solubility Information H2O : 100 mg/mL (347.05 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (6.94 mM; Need ultrasonic)
SMILES OC(C1=CC(CO)=C(O)C=C1)CNC(C)(C)C.[0.5H2SO4]
Molecular Weight (g/mol) 288.14
Formula Weight 288.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only