missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salbutamol Recombinant Rabbit Monoclonal Antibody (Huam003-82-2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA545006
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.
Specifications
| Salbutamol | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS, TBS with 0.05% sodium azide; pH 7.4 | |
| Salbutamol linked to BSA. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
| ELISA | |
| Huam003-82-2 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction