missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salbutamol Sulphate (Albuterol Sulphate) 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Salbutamol Sulphate (Albuterol Sulphate) 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Salbutamol sulfate |
| CAS | 51022-70-9 |
| Analyte or Component Names | Salbutamol sulfate |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | 2 C13 H21 N O3 . H2 O4 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Salbutamol Sulphate, Albuterol Sulphate, AH 3365 F |
| InChI Formula | InChI=1 S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17 H,7-8H2,1-3H3;(H2,1,2,3,4) |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2.OS(=O)(=O)O |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction