missing translation for 'onlineSavingsMsg'
Learn More

Salicin, MedChemExpress

Product Code. 16422356
Encompass
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422356 Supplier Medchem Express Supplier No. HYN0149

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Salicin is a natural COX inhibitor.

Salicin is a natural COX inhibitor.

Specifications

Chemical Name or Material Salicin
CAS 138-52-3
Color White
Purity Grade Notes Research
Molecular Formula C13H18O7
Quantity 500 mg
Synonym D-(−)-Salicin Salicoside
Solubility Information DMSO : 150 mg/mL (523.96 mM; Need ultrasonic and warming)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OC2=CC=CC=C2CO
Molecular Weight (g/mol) 286.28
Formula Weight 286.28
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.