missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Salicylic acid-D4 |
| CAS | 78646-17-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 2H4 H2 O3 |
| Quantity | 5 mg |
| Synonym | Benzoic-2,3,4,5-d4 acid','6-hydroxy-, 6-Hydroxybenzoic-2,3,4,5-d4 acid, 2H4-Salicylic acid, Salicylic acid-D4 (phenyl-D4), 2-Carboxyphenol-3,4,5,6-d4, 2-Hydroxybenzene-3,4,5,6-d4-carboxylic acid, o-Carboxyphenol-d4, o-Hydroxybenzoic-d4 acid, 2-Hydroxy-3,4,5,6-tetradeuteriobenzoic acid |
| InChI Formula | InChI=1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)O)c(O)c1[2H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 142.15 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?