Learn More
Salicylideneaniline, 97%, Thermo Scientific™
Product Code. 11329976 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
Product Code. 11329976
Brand: Thermo Scientific Acros 338980250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 779-84-0
Melting Point 49.4°C to 52.4°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C13H11NO
Linear Formula C6H5NCHC6H4(OH)
Quantity 25 g
Synonym n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, chloroform, slightly soluble in hexane, methanol
InChI Key GVHXCXKLYZOBKS-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC=C2C=CC=CC2=O
IUPAC Name 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol) 197.24
PubChem CID 6740751
Formula Weight 197.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Salicylideneaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Salicylideneaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.