Learn More
Salicylideneaniline, 98%
Product Code. 11497006
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11497006
Brand: Thermo Scientific Alfa Aesar B25332.09
Request Bulk or Custom Format
CAS: 779-84-0 | C13H11NO | 197.237 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 779-84-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00020066
InChI Key GVHXCXKLYZOBKS-UHFFFAOYSA-N
Synonym n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl
PubChem CID 6740751
IUPAC Name 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC=C(C=C1)NC=C2C=CC=CC2=O
Specifications
Melting Point 48°C to 52°C
Boiling Point 151°C (2 mmHg)
Quantity 10 g
Beilstein 1367639
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material Salicylideneaniline
compliance-icons
Product Identifier
  • Salicylideneaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Salicylideneaniline