missing translation for 'onlineSavingsMsg'
Learn More

Salidroside, TRC

Product Code. 30011484
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011484 50 mg 50mg
30003565 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011484 Supplier Toronto Research Chemicals Supplier No. TRCS08860050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10338-51-9
Molecular Formula C14 H20 O7
Molecular Weight (g/mol) 300.3
Synonym Salidroside, Sallidroside, Tyrosol alpha-(beta-D-glucopyranoside), Rhodioloside, Rhodosin, 4-Hydroxy-phenyl-2-ethyl-beta-D-glucopyranoside, 2-(4-Hydroxyphenyl)ethyl-beta-D-glucopyranoside, p-Hydroxyphenethyl Glucopyranoside, 2-(4-Hydroxyphenyl)ethyl beta-D-Glucopyranoside, 2-(4-Hydroxyphenyl)ethyl-β-D-glucopyranoside, 4-Hydroxy-phenyl-2-ethyl-β-D-glucopyranoside, Tyrosol α-(β-D-Glucopyranoside)
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[2-(4-hydroxyphenyl)ethoxy]oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OCCc2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H20O7/c15-7-10-11(17)12(18)13(19)14(21-10)20-6-5-8-1-3-9(16)4-2-8/h1-4,10-19H,5-7H2/t10-,11-,12+,13-,14-/m1/s1
Formula Weight 300.1209
Chemical Name or Material Salidroside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.