Learn More
SALL4 Antibody (CL12185) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15130JF525
Product Code. 30288216
  • € 737.00 / 0.10mL
Description
SALL4 Monoclonal antibody specifically detects SALL4 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SALL4
Monoclonal
Janelia Fluor 525
dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797sal (Drosophila)-like 4
This antibody was developed using a synthetic peptide derived from Q9UJQ4, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunocytochemistry/Immunofluorescence
CL12185
50mM Sodium Borate
Mouse
Protein A purified
RUO
57167
Store at 4⪚C in the dark.