missing translation for 'onlineSavingsMsg'
Learn More

SALL4 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. 16842975 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16842975 20 μL 20µL
16832975 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16842975 Supplier Proteintech Supplier No. 245001AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sall4 is a probable transcription factor. Defects in Sall4 are the cause of Okihiro syndrome; also known as Duane radial ray syndrome (DRRS). It is a disorder characterized by the association of forearm malformations with Duane retraction syndrome. Sall4 is also involved in forelimb and heart development in mice.

Specifications

Antigen SALL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SALL4
Gene Accession No. Q9UJQ4
Gene Alias DRRS, HSAL4, sal like 4 (Drosophila), Sal like protein 4, SALL4, Zinc finger protein SALL4, ZNF797
Gene Symbols SALL4
Host Species Rabbit
Immunogen SALL4 Fusion Protein Ag17480
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57167, 686412
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.