missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella Antibody (100/358), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64563UV
This item is not returnable.
View return policy
Description
Salmonella Monoclonal specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Salmonella | |
| Monoclonal | |
| DyLight 350 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Antibody recognizes an antigen common to the various Salmonella strains. | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 100/358 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Native S. enteritidis (NCTC 5188 9.12:g,m). | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction