missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella Antibody (100/358), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64563JF646
This item is not returnable.
View return policy
Description
Salmonella Monoclonal specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Salmonella | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Native S. enteritidis (NCTC 5188 9.12:g,m). | |
| 0.1 mL | |
| Antibody recognizes an antigen common to the various Salmonella strains. | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 100/358 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction