missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salmonella paratyphi C1 LPS Monoclonal Antibody (6344)

Description
Salmonella paratyphi C1 LPS Monoclonal Antibody for ICC/IF, ELISA
Salmonella paratyphi C1 LPS (lipopolysaccharide) is a complex molecule found on the surface of the bacterium Salmonella paratyphi C1. It is composed of a lipid portion and a polysaccharide portion, and plays a crucial role in the pathogenesis of Salmonella infections. The LPS molecule is recognized by the host immune system and triggers an inflammatory response, leading to the symptoms of salmonellosis.
Specifications
Specifications
| Antigen | Salmonella paratyphi C1 LPS |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6344 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Salmonellae |
| Host Species | Mouse |
| Purification Method | Protein A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?