missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salmonella typhi Monoclonal Antibody (AC04)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546571
This item is not returnable.
View return policy
Description
Salmonella typhi Monoclonal Antibody for ICC/IF, ELISA
A ubiquitous human and animal pathogen, salmonella is the causative agent of salmonellosis, a disease which is common throughout the world. The disease is usually not severe in humans, but can occasionally cause serious systemic infections which require antibiotic treatment. As salmonellosis is a zoonosis with a large animal reservoir, contaminated food is by far the most common mode of transmission of non-typhoidal salmonellae. The large and varied animal reservoir of salmonella has made control of salmonellosis a difficult task. While an effective vaccine does exist against typhoid fever, there is none against non-typhoidal salmonellosis.
Specifications
| Salmonella typhi | |
| Monoclonal | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| AC04 | |
| Unconjugated | |
| S. typhi | |
| Native Salmonella typhi. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction