missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella typhimurium Antibody (1E6cc), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-16952JF549
This item is not returnable.
View return policy
Description
Salmonella typhimurium Monoclonal antibody specifically detects Salmonella typhimurium in Bacteria samples. It is validated for ELISA
Specifications
| Salmonella typhimurium | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| 1E6cc | |
| ELISA | |
| S. typhimurium;Salmonella LPS;Salmonellae | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Lipopolysaccharides (LPS) of S. typhimurium. | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction