missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD12 Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809016S
This item is not returnable.
View return policy
Description
SAMD12 Monoclonal antibody specifically detects SAMD12 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SAMD12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78352 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1742 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| DLG4 | |
| SAM domain-containing protein 12; SAMD12; sterile alpha motif domain containing 12; sterile alpha motif domain-containing protein 12 | |
| Full length human recombinant protein of human DLG4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction