missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LKSLRLHKYA ALFSQMTYEE MMALTECQLE AQNVTKGARH KIVISIQKLK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
Sterile alpha motifs (SAMs) in proteins such as SAMD4A are part of an RNA-binding domain that functions as a posttranscriptional regulator by binding to an RNA sequence motif known as the Smaug recognition element, which was named after the Drosophila Smaug protein.
Specifications
Specifications
| Antigen | SAMD4A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Samd4a |
| Gene Accession No. | Q9UPU9 |
| Gene Alias | 1700024G08Rik; 1700111L17Rik; 4933436G17Rik; KIAA1053; Protein Smaug homolog 1; SAM domain-containing protein 4A; Samd4; SAMD4A; Smaug; smaug 1; Smaug1; SMG; SMGA; sterile alpha motif domain containing 4; sterile alpha motif domain containing 4A; sterile alpha motif domain-containing protein 4A |
| Gene Symbols | Samd4a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
