missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMHD1 Recombinant Rabbit Monoclonal Antibody (24GB1010)

Rabbit Recombinant Monoclonal Antibody
€ 204.00 - € 580.00
Specifications
| Antigen | SAMHD1 |
|---|---|
| Clone | 24GB1010 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275694
![]() |
Invitrogen™
MA557700 |
100 μL |
€ 580.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276397
|
Invitrogen™
MA557701 |
20 μL |
€ 204.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SAMHD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.Specifications
| SAMHD1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| SAMHD1 | |
| CHBL2; DCIP; Dendritic cell-derived IFNG-induced protein; deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; E330031J07Rik; EGK_02438; HDDC1; hSAMHD1; IFN-gamma induced; interferon-gamma-inducible protein Mg11; Mg11; Mg21; Monocyte protein 5; MOP5; MOP-5; mSAMHD1; RP1-132F21.1; SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1; SAM domain and HD domain 1; SAM domain and HD domain, 1; SAM domain and HD domain-containing protein 1; Samhd1; SBBI88 | |
| SAMHD1 | |
| Primary | |
| Affinity chromatography |
| 24GB1010 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y3Z3 | |
| 25939 | |
| A synthesized peptide derived from human SAMHD1 (aa 580-626). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title