missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMM50 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556969
This item is not returnable.
View return policy
Description
Immunogen sequence: HARSLEPLPS SGPDFGGLGE EAEFVEVEPE AKQEILENKD VVVQHVHFDG LGRTKDDIII CEIGDVFKAK NLIEVMRKSH EAREKLLRLG IFRQVDVLID Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications
| SAMM50 | |
| Polyclonal | |
| Unconjugated | |
| Samm50 | |
| 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25813, 300111, 68653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6AXV4, Q8BGH2, Q9Y512 | |
| Samm50 | |
| Recombinant protein corresponding to Human SAMM50. Recombinant protein control fragment (Product #RP-95359). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction