missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMM50 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118251
This item is not returnable.
View return policy
Description
SAMM50 Polyclonal Antibody for Western Blot, IHC (P), IP
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications
| SAMM50 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9Y512 | |
| Samm50 | |
| E. coli-derived Human SAMM50 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Samm50 | |
| 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 25813 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction