Learn More
Invitrogen™ SAMM50 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531202
Description
Recommended positive controls: A431 cytosol. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (85%), Xenopus laevis (88%), Chicken (92%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications
| SAMM50 | |
| Polyclonal | |
| Unconjugated | |
| Samm50 | |
| 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25813, 68653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.83 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8BGH2, Q9Y512 | |
| Samm50 | |
| Recombinant protein encompassing a sequence within the center region of human SAMM50. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.