Learn More
Rheodyne Sample loop (9725, 9010, pr/ev750-100, pr/ev753-105) PEEK(TM) 1/16 inch O.D., 0.020inch (0.51mm)
Brand:  Rheodyne 02016-80
Sample loop (9725, 9010, pr/ev750-100, pr/ev753-105) PEEK(TM) 1/16 inch O.D., 0.020inch (0.51mm)
Product Code. 12926109
  • ONLINE EXCLUSIVE PRICE
    € 167.00
    € 132.46 / Each
    Save € 34.54
    20% Off