Learn More
Cole-Parmer Sanitary Clamp Fittings, 90 Degrees Elbow Union, 316 Stainless Steel
Product Code. 30053695 Shop All Cole-Parmer Products
Click to view available options
Includes:
1 in. Sanitary Clamp Size
1-1/2 in. Sanitary Clamp Size
2 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Unit Size:
Each
Product Code. 30053695
Brand: Cole-Parmer 5011828
Only null left
Add to basket
Add to basket
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
Specifications
Certifications/Compliance 3 A;FDA
Outlet Port Sanitary
Shape 90° Elbow
Includes 1 in. Sanitary Clamp Size