Learn More
Cole-Parmer Sanitary Clamp Fittings, Straight Eccentric Reducer, 316 Stainless Steel
Product Code. p-200105210 Shop All Cole-Parmer Products
Click to view available options
Includes:
1 to 3/4 in. Sanitary Clamp Size
1-1/2 to 1 in. Sanitary Clamp Size
2 to 1 in. Sanitary Clamp Size
Unit Size:
Each
Product Code. 30054084
Brand: Cole-Parmer 5011804
Only null left
Add to basket
Add to basket
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases. Offset centerline—useful when both air and liquids are flowing together
Specifications
Certifications/Compliance 3 A;FDA
Outlet Port Sanitary
Shape Straight
Includes 1 to 3/4 in. Sanitary Clamp Size