Learn More
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone
Product Code. 12479650
Click to view available options
Tubing Size:
Fits 1/2 in. ID tubing
Fits 3/8 in. ID tubing
Unit Size:
Each
Product Code. 12479650
Brand: Cole-Parmer™ 3151208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone has a tapered funnel design to provide a steady fluid flow.
  • Reduces dead space and improves flow.
  • Tapered funnel design provides a steady fluid flow.
  • Smooth inner surface prevents particle entrapment.
  • Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
  • Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Certifications/Compliance Meets USP Class VI requirements and is FDA compliant for food contact
Autoclavable Not Autoclavable
Material Polysulfone
For Use With 1/2 in. ID tubing, 1/2 in. mini clamp
For Use With (Application) FDA compliant for food contact
Tubing Size Fits 1/2 in. ID tubing
Sterilization Method Gamma
Width (English) 31/32 in.
Height (English) 1 13/16 in.