Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Record Tip
Poulten & Graf GmbH FORTUNA™ OPTIMA™ Glass and Metal Syringe with Record Tip is used to measure and dispense fluids.
Brand: Poulten & Graf GmbH 7 20041
Additional Details : Weight : 0.10000kg
Description
- With recorder tip
- With steel piston
- Interchangable
- For technical use only
Specifications
Autoclavable | |
Clear | |
Yes | |
No | |
Glass and Metal Syringe | |
20 mL | |
Yes |
Steel (Plunger), Nickel-plated Brass (Tip), Neutral Glass (Barrel) | |
Glass and Metal Syringe with Record tip | |
Glass and Metal Syringe | |
Amber | |
1 | |
Record Tip |