missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAP (SLAM-Associated Protein) Monoclonal Antibody (10C4.2), eBioscience™

Product Code. p-7084913
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15278207 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15278207 Supplier Invitrogen™ Supplier No. 14988882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: This 10C4.2 monoclonal antibody reacts with human SLAM-associated protein (SAP). Upon SLAM receptor ligation, this 16-kDa SH2 domain-containing adaptor protein recruits Fyn tyrosine kinase, which phosphorylates the receptor to activate downstream signaling events. SAP is expressed in T, NK, and NKT cells, as well as eosinophils and platelets. This protein has also been detected in B cells and tumor cell lines such as Jurkat and Raji. Studies demonstrate that SAP expression in T follicular helper cells is required for germinal center formation. Moreover, this protein is involved in NKT cell development, Th1/Th2 differentiation, and controlling NK cytotoxicity. The role of SAP in B cells, however, is currently under investigation. Finally, deletion of the gene encoding SAP, SH2D1A, results in X-linked lymphoproliferative disease in humans. Applications Reported: This 10C4-SAP (10C4.2) antibody has been reported for use in immunoblotting and immunocytochemistry (ICC). Applications Tested: This 10C4.2 antibody has been tested by immunoblotting of reduced cell lysate prepared from Jurkat cells. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP (SLAM-Associated Protein)
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10C4.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Sh2d1a
Gene Accession No. O60880
Gene Alias DSHP; Duncan disease homolog; Duncan disease SH2-protein; Duncan's disease; EBVS; FLJ18687; FLJ92177; Gm686; HGNC:10820; IMD5; LYP; MTCP1; RGD1562408; RP5-1052M9.3; SAP; SAP/SH2D1A; SH2 domain containing 1A; SH2 domain protein 1A; SH2 domain-containing protein 1A; SH2D1A; signaling lymphocyte activation molecule-associated protein; signaling lymphocytic activation molecule-associated protein; SLAM associated protein/SH2 domain protein 1A; SLAM-associated protein; SLAM-associated protein isoform SAP-2; T cell signal transduction molecule SAP; T-cell signal transduction molecule SAP; Xlp; XLPD; XLPD1
Gene Symbols Sh2d1a
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4068
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.