missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP102 Antibody (OTI3F2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73985F
This item is not returnable.
View return policy
Description
SAP102 Monoclonal specifically detects SAP102 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| SAP102 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| DLG3 | |
| Full length recombinant protein of human SAP102 (NP_066943) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI3F2 | |
| Flow Cytometry | |
| discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Neuroscience | |
| 1741.0 | |
| Store at 4°C in the dark. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering