missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP30 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92648AF594
This item is not returnable.
View return policy
Description
SAP30 Polyclonal antibody specifically detects SAP30 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SAP30 | |
| Polyclonal | |
| Western Blot | |
| 30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa | |
| The immunogen this antibody was made to, maps to a region between residue 170 and 220 of human Sin3A-Associated Protein, 30kDa using the numbering given in entry NP_003855.1 (GeneID 8819). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8819 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction