missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAP30BP Monoclonal Antibody (OTI4H1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526587
This item is not returnable.
View return policy
Description
SAP30BP Monoclonal Antibody for Western Blot, IHC (P)
Induces cell death. May act as a transcriptional corepressor of a gene related to cell survival. May be involved in the regulation of beta-2-microglobulin genes. [UniProt].
Specifications
| SAP30BP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHR5 | |
| Sap30bp | |
| Human recombinant protein fragment corresponding to amino acids 1-260 of SAP30BP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H1 | |
| Unconjugated | |
| Sap30bp | |
| 2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29115 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction