missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP30BP Monoclonal Antibody (OTI4H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803237S
This item is not returnable.
View return policy
Description
SAP30BP Monoclonal antibody specifically detects SAP30BP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SAP30BP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09327 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7429 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H1 | |
| Unconjugated | |
| VIL1 | |
| 2700016D05Rik; fb33d03; Hcngp; HSV-1 binding; HTRG; HTRP; RGD1565537; SAP30 binding protein; SAP30-binding protein; Sap30bp; transcriptional regulator protein; transcriptional regulator protein (Hcngp); transcriptional regulator protein HCNGP; Unknown (protein for MGC:142452); wu:fb33d03; zgc:55340 | |
| Human recombinant protein fragment corresponding to amino acids 1-188 of human VIL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction