Learn More
Invitrogen™ SAPAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139783
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PKTSPKAVAR RFTTRRSSSV DQARINCCVP PRIHPRSSIP GYSRSLTTGQ LSDELNQQLE AVCGSVFGEL ES Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.
SAP90/PSD-95-associated protein 3 (SAPAP3, also known as DLGAP3) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP3 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). Both SAPAP3 protein and mRNA are targeted to dendrites, whereas SAPAP1, -2, and -4 mRNAs are detected mainly in cell bodies. Recent experiments have suggested that SAPAP3 may be involved in obsessive-compulsive disorder (OCD), as mice lacking SAPAP3 exhibited OCD-like symptoms which could be relieved by lentiviral-mediated selective expression of SAPAP3 in the striatum of SAPAP3-mutant mice. At least two isoforms are known to exist.
Specifications
| SAPAP3 | |
| Polyclonal | |
| Unconjugated | |
| DLGAP3 | |
| BC058433; DAP3; DAP-3; discs large homolog associated protein 3; discs, large (Drosophila) homolog-associated protein 3; disks large-associated protein 3; DLG associated protein 3; Dlgap3; Prpl8; PSD-95/SAP90-associated protein-3; PSD-95/SAP90-binding protein 3; SAP90/PSD 95 associated protein 3; SAP90/PSD-95-associated protein 3; SAPAP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95886 | |
| DLGAP3 | |
| Recombinant protein corresponding to Human DLGAP3. Recombinant protein control fragment (Product #RP-108767). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.