Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Attached Cap
Product Code. 13022023
Change view
Click to view available options
Volume (Metric):
0.2 mL
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
13022023 0.2 mL Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13022023 Supplier Greiner Bio-One Supplier No. 608281
Add to basket
Edge
Add to basket
Greiner Bio-One Sapphire PCR 8-Tube Strip with individually attached cap is thin wall for optimal heat transfer.
  • Non-pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
  • Individually attached caps to minimise cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 0.2 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap (Attached)
Product Type PCR 8-Tube Strip