Learn More
Saquinavir Mesylate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Saquinavir mesylate is an HIV Protease Inhibitor used in antiretroviral therapy.
Specifications
Specifications
| Chemical Name or Material | Saquinavir Mesylate |
| CAS | 149845-06-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C39H54N6O8S |
| Quantity | 10 mg |
| Synonym | Ro 31-8959/003 |
| Solubility Information | DMSO : 20 mg/mL (26.08 mM; Need ultrasonic) |
| SMILES | NC(C[C@H](C(N[C@@H](CC1=CC=CC=C1)C(CN2C[C@@]3([H])CCCC[C@]3(C[C@H]2C(NC(C)(C)C)=O)[H])O)=O)NC(C4=NC5=CC=CC=C5C=C4)=O)=O.CS(O)(=O)=O |
| Molecular Weight (g/mol) | 766.95 |
| Show More |
Research purposes only