missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR1B Antibody (1C7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22566CL1
This item is not returnable.
View return policy
Description
SAR1B Monoclonal antibody specifically detects SAR1B in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SAR1B | |
| Monoclonal | |
| CoraFluor 1 | |
| 2310075M17Rik, ANDD, CMRD, EC 3.6.5, GTP-binding protein B, GTP-binding protein SAR1b, GTP-binding protein Sara, SAR1 homolog B (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 2, SAR1a gene homolog 2, SAR1a gene homolog 2 (S. cerevisiae), SARA2GTBPB, SARB | |
| Recombinant human SAR1B (1-198aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 51128 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1C7 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction