missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33521
This item is not returnable.
View return policy
Description
SAR1B Polyclonal specifically detects SAR1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SAR1B | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9Y6B6 | |
| SAR1B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TLLHMLKDDRLGQHVPTLHPTSEELTIAGMTFTTFDLGGHVQARRVWKNYLPAINGIVFLVDCADHE | |
| 0.1 mL | |
| Signal Transduction | |
| 51128 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2310075M17Rik, ANDD, CMRD, EC 3.6.5, GTP-binding protein B, GTP-binding protein SAR1b, GTP-binding protein Sara, SAR1 homolog B (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 2, SAR1a gene homolog 2, SAR1a gene homolog 2 (S. cerevisiae), SARA2GTBPB, SARB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction