missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sarcalumenin Antibody (XIIC4), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2874JF549
This item is not returnable.
View return policy
Description
Sarcalumenin Monoclonal antibody specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot
Specifications
| Sarcalumenin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| sarcalumenin | |
| Purified rabbit sarcoplasmic reticulum membrane vesicles. | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 6345 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| XIIC4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse, Canine, Guinea Pig, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction