missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARG Polyclonal Antibody

Product Code. 15710821 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15710821 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710821 Supplier Invitrogen™ Supplier No. PA552868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SGEDPNSRLA PLTTPKPRKL PPNIVLKSSR SSFHSDPQHW LSRHTEAAPG DSGLISCSLQ EQRKARKEAL EKLGLPQDQD EPGLHLSKPT SSIRPKETRA QHLSPAPGLA QPAAPAQASA AIPAAG Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 61%.

Specifically androgen-regulated gene protein (SARG), encoded by C1orf116, is a 601-aa protein considered putative androgen-specific receptor. SARG mRNA is highly expressed in prostate and can be up-regulated by androgens, but not by glucocorticoids. Little is known about the function of SARG to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1orf116
Gene Accession No. Q9BW04
Gene Alias AA986860; C1orf116; chromosome 1 open reading frame 116; E030025M07; expressed sequence AA986860; Sarg; Specifically androgen-regulated gene protein; specifically androgen-regulated protein
Gene Symbols C1orf116
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SARG. Recombinant protein control fragment (Product #RP-89672).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.