missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SARM1 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 23098 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-29625. For further blocking peptide related protocol, click here . |
| Gene Alias | KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog |
| Gene Symbol | SARM1 |
| Quantity | 0.1 mg |
| Research Category | Innate Immunity |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction