missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS/SARS-CoV-2 Nucleocapsid Monoclonal Antibody (E16C)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA17403

Product Code. 11520542

  • € 534.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-7403 is reactive with the NP of SARS-CoV-2 (COVID-19 virus) by ELISA. Negative for human coronaviruses 229E & OC43. ELISA dilution range suggested 1:20- 1:200. The MA1-7403 antibody reacts with SARS Coronavirus nucleoprotein in viral and infected tissue samples. MA1-7403 has been successfully used in ELISA, Western blot and Immunofluorescence applications. The MA1-7403 antibody was raised against a SARS virus nucleoprotein preparation.

SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SARS/SARS-CoV-2 Nucleocapsid
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
P0DTC9
N
SARS nucleoprotein preparation.
100 μg
Primary
Virus
Antibody
IgG2b
ELISA, Immunohistochemistry, Western Blot
E16C
Unconjugated
N
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
Mouse
Protein A
RUO
43740575
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.