Learn More
SARS-CoV-2 3CL Protease Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-07062PCP
This item is not returnable.
View return policy
Description
SARS-CoV-2 3CL Protease Polyclonal antibody specifically detects SARS-CoV-2 3CL Protease in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Western Blot, Immunohistochemistry | |
| PerCP | |
| Rabbit | |
| 43740578 |