Learn More
SARS-CoV-2 Membrane Protein Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-07058MFV610
Product Code. 18577210
  • € 666.00 / 0.10mL
Description
SARS-CoV-2 Membrane Protein Polyclonal antibody specifically detects SARS-CoV-2 Membrane Protein in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunohistochemistry
Affinity purified
IgG
Western Blot, ELISA, Immunohistochemistry
Store at 4C in the dark.