missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 N Protein Mouse anti-Virus, Clone: OTI10A8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814447S
This item is not returnable.
View return policy
Description
SARS-CoV-2 N Protein Monoclonal antibody specifically detects SARS-CoV-2 N Protein in Virus samples. It is validated for ELISASpecifications
| SARS-CoV-2 N Protein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P59595 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1489678 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI10A8 | |
| Unconjugated | |
| N | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid | |
| Full length recombinant SARS-Cov-2 N protein produced in E.coli. | |
| 30 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction