Learn More
Invitrogen™ SARS-CoV-2 NSP11 (strain Wuhan-Hu-1) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143475
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The SARS-CoV-2 has a large proteome, in which, structure and functions of all proteins are not known yet, along with non-structural protein 11 (nsp11) In the presence of negatively charged and neutral liposomes, nsp11 remains disordered However, with SDS micelle, it adopted an alpha-helical conformation, suggesting the helical propensity of nsp11 [PMID: 34119626].
Specifications
| SARS-CoV-2 NSP11 (strain Wuhan-Hu-1) | |
| Polyclonal | |
| Unconjugated | |
| CoV-2; NSP11 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of SARS-CoV-2 (COVID-19) Nsp11 (SARS-CoV-2 (strain Wuhan-Hu-1)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |