Learn More
Invitrogen™ SARS-CoV-2 NSP11 (strain Wuhan-Hu-1) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5143475
Product Code. 17890636
  • € 455.00 / 100µL
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The SARS-CoV-2 has a large proteome, in which, structure and functions of all proteins are not known yet, along with non-structural protein 11 (nsp11) In the presence of negatively charged and neutral liposomes, nsp11 remains disordered However, with SDS micelle, it adopted an alpha-helical conformation, suggesting the helical propensity of nsp11 [PMID: 34119626].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 NSP11 (strain Wuhan-Hu-1)
Polyclonal
Unconjugated
CoV-2; NSP11
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of SARS-CoV-2 (COVID-19) Nsp11 (SARS-CoV-2 (strain Wuhan-Hu-1)). The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Rabbit
Protein A
RUO
Virus