missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP12 Recombinant Rabbit Monoclonal Antibody (HL1444)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547047
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Responsible for replication and transcription of the viral RNA genome.
Specifications
| SARS-CoV-2 NSP12 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL1444 | |
| Unconjugated | |
| COVID19 Nsp12; COVID-19 Nsp12; SARS-CoV-2 Nonstructural Protein 12; SARS-CoV2 NSP12; SARS-COV-2 NSP12 | |
| Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp12 / RdRP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction