missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143484
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Responsible for replication and transcription of the viral RNA genome.
Specifications
| SARS-CoV-2 NSP12 | |
| Polyclonal | |
| Unconjugated | |
| COVID19 Nsp12; COVID-19 Nsp12; SARS-CoV-2 Nonstructural Protein 12; SARS-CoV2 NSP12; SARS-COV-2 NSP12 | |
| Recombinant protein encompassing a sequence within the RdRP domain of SARS-CoV-2 (COVID-19) nsp12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction