Learn More
SARS-CoV-2 nsp13 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-07056MFV610
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp13 Polyclonal antibody specifically detects SARS-CoV-2 nsp13 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry
Specifications
| SARS-CoV-2 nsp13 | |
| Polyclonal | |
| ELISA, Immunohistochemistry | |
| helicase, Non-structural protein 13 | |
| Purified |
| ELISA, Immunohistochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Store at 4C in the dark. |