missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 NSP13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17921051
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921051 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921051 Supplier Invitrogen™ Supplier No. PA5143816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted reactivity based on immunogen sequence: SARS-CoV NSP13 (Helicase) protein: (80%).

SARS-CoV-2 NSP13, also known as helicase, is another essential component of the coronavirus responsible for the COVID-19 pandemic. It plays a crucial role in viral replication and transcription by unwinding double-stranded RNA, facilitating the separation of RNA strands during replication. NSP13 also interacts with other proteins in the viral replication complex, making it a potential target for antiviral drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP13
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ORF1a
Gene Accession No. P0C6V0
Gene Symbols ORF1a
Host Species Rabbit
Immunogen A peptide corresponding to 19 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP13 (Helicase) protein. The immunogen is located in the last 50 amino acids of SARS-CoV-2 (COVID-19) NSP13 (Helicase).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489749
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.