missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 nsp15 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11932MFV610
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp15 Polyclonal antibody specifically detects SARS-CoV-2 nsp15 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Specifications
| SARS-CoV-2 nsp15 | |
| Polyclonal | |
| Western Blot, ELISA | |
| endoRNAse, Non-structural protein 15 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxyl terminus of SARS-CoV-2 NSP15 protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 NSP15 protein. | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction