missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 NSP8 Recombinant Rabbit Monoclonal Antibody (HL1523)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA547079

Product Code. 17829255

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The nonstructural protein 8 (nsp8) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp8 is thought to be part of the viral replication complex, which is associated with intracellular membranes. No specific information on the function of nsp8 is available.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SARS-CoV-2 NSP8
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL1523
Unconjugated
CoV-2; non-structural protein 8; NSP8
Recombinant protein encompassing a sequence within the N-terminus region of SARS-CoV-2 (COVID-19) nsp8. The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.